Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: didum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057838.4 | NP_477186.1 | 11 - 12 | 3994 | 85 | |
NM_165535.2 | NP_724569.1 | 12 - 13 | 4018 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |