Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: didum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057838.4 | NP_477186.1 | 3 - 4 | 1421 | 67 | |
NM_165535.2 | NP_724569.1 | 3 - 4 | 1421 | 67 | ||
NM_165536.3 | NP_724570.1 | 3 - 4 | 1421 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |