Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup44A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136499.3 | NP_610343.1 | 2 - 3 | 646 | 105 | |
NM_165582.2 | NP_724637.1 | 3 - 4 | 721 | 105 | ||
NM_165583.2 | NP_724638.1 | 3 - 4 | 769 | 105 | ||
GenBank mRNA | AY060480.1 | - | 2 - 3 | 1850 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |