Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057554.4 | NP_476902.1 | 2 - 3 | 508 | 91 | |
NM_165589.3 | NP_724648.1 | 2 - 3 | 508 | 91 | ||
GenBank mRNA | AY070631.1 | - | 2 - 3 | 506 | 91 | |
AY075519.1 | - | 2 - 3 | 506 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |