Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165622.2 | NP_724708.1 | 3 - 4 | 1316 | 91 | |
GenBank mRNA | AY061319.1 | - | 3 - 4 | 1293 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |