Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014513.2 | NP_001014513.1 | 3 - 4 | 875 | 72 | |
NM_001299282.1 | NP_001286211.1 | 3 - 4 | 497 | 72 | ||
NM_001382198.1 | NP_001369066.1 | 2 - 3 | 918 | 72 | ||
NM_001382199.1 | NP_001369067.1 | 2 - 3 | 918 | 72 | ||
NM_165624.2 | NP_724710.1 | 3 - 4 | 875 | 72 | ||
NM_165625.2 | NP_724711.1 | 3 - 4 | 497 | 72 | ||
GenBank mRNA | AY061062.1 | - | 3 - 4 | 493 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |