Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014513.2 | NP_001014513.1 | 6 - 7 | 1292 | 100 | |
NM_001299282.1 | NP_001286211.1 | 6 - 7 | 914 | 100 | ||
NM_001382198.1 | NP_001369066.1 | 4 - 5 | 1293 | 100 | ||
NM_001382199.1 | NP_001369067.1 | 5 - 6 | 1335 | 100 | ||
NM_165624.2 | NP_724710.1 | 5 - 6 | 1250 | 100 | ||
NM_165625.2 | NP_724711.1 | 5 - 6 | 872 | 100 | ||
GenBank mRNA | AY061062.1 | - | 5 - 6 | 868 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |