Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014513.2 | NP_001014513.1 | 8 - 9 | 2779 | 60 | |
NM_001299282.1 | NP_001286211.1 | 8 - 9 | 2401 | 60 | ||
NM_001382198.1 | NP_001369066.1 | 6 - 7 | 2780 | 60 | ||
NM_001382199.1 | NP_001369067.1 | 7 - 8 | 2822 | 60 | ||
NM_165624.2 | NP_724710.1 | 7 - 8 | 2737 | 60 | ||
NM_165625.2 | NP_724711.1 | 7 - 8 | 2359 | 60 | ||
GenBank mRNA | AY061062.1 | - | 7 - 8 | 2355 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |