Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014514.2 | NP_001014514.1 | 6 - 7 | 2348 | 125 | |
NM_080371.3 | NP_525110.2 | 5 - 6 | 2273 | 125 | ||
NM_165666.2 | NP_724772.1 | 5 - 6 | 2273 | 125 | ||
NM_165667.2 | NP_724773.1 | 6 - 7 | 2348 | 125 | ||
GenBank mRNA | AY069188.1 | - | 5 - 6 | 2257 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |