Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259296.2 | NP_001246225.1 | 4 - 5 | 1253 | 60 | |
NM_001259297.2 | NP_001246226.2 | 4 - 5 | 1363 | 60 | ||
NM_136644.3 | NP_610488.1 | 4 - 5 | 1048 | 60 | ||
NM_165675.3 | NP_724789.1 | 4 - 5 | 1173 | 60 | ||
NM_165676.2 | NP_724790.1 | 4 - 5 | 1363 | 60 | ||
GenBank mRNA | AF204777.1 | - | 4 - 5 | 1082 | 60 | |
AY119596.1 | - | 4 - 5 | 1081 | 60 | ||
BT125651.1 | - | 4 - 5 | 1254 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |