Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG46338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273908.1 | NP_001260837.1 | 2 - 3 | 128 | 72 | |
NM_080149.4 | NP_524888.1 | 1 - 2 | 177 | 72 | ||
NM_165716.2 | NP_724848.1 | 2 - 3 | 305 | 72 | ||
GenBank mRNA | AY051650.1 | - | 1 - 2 | 158 | 72 | |
BT001497.1 | - | 2 - 3 | 305 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |