Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp33Eb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135726.3 | NP_609570.1 | 2 - 3 | 441 | 76 | |
GenBank mRNA | AY071008.1 | - | 2 - 3 | 439 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |