Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136821.3 | NP_610665.2 | 2 - 3 | 1614 | 79 | |
NM_165815.2 | NP_725019.1 | 2 - 3 | 1289 | 79 | ||
NM_165816.2 | NP_725020.1 | 2 - 3 | 1012 | 79 | ||
GenBank mRNA | BT023808.1 | - | 2 - 3 | 1265 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |