Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144691.2 | NP_001138163.1 | 4 - 5 | 595 | 89 | |
NM_001272337.1 | NP_001259266.1 | 4 - 5 | 595 | 89 | ||
NM_078499.5 | NP_511054.4 | 4 - 5 | 740 | 89 | ||
GenBank mRNA | BT120367.1 | - | 4 - 5 | 742 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |