Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078500.4 | NP_511055.1 | 4 - 5 | 1380 | 57 | |
GenBank mRNA | AY095178.1 | - | 4 - 5 | 1297 | 57 |
Gene Symbol: raptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297995.1 | NP_001284924.1 | 4 - 5 | 5944 | 57 | |
NM_132066.2 | NP_572294.2 | 4 - 5 | 5944 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |