Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: org-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298090.1 | NP_001285019.1 | 1 - 2 | 942 | 76 | |
NM_078530.3 | NP_511085.2 | 1 - 2 | 742 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |