Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ant2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272503.1 | NP_001259432.1 | 2 - 3 | 396 | 78 | |
NM_078555.4 | NP_511110.1 | 1 - 2 | 465 | 78 | ||
NM_176725.3 | NP_788898.1 | 2 - 3 | 432 | 78 | ||
GenBank mRNA | BT044582.1 | - | 1 - 2 | 440 | 78 | |
BT044583.1 | - | 1 - 2 | 442 | 78 | ||
BT088432.1 | - | 2 - 3 | 405 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |