Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078575.3 | NP_511130.2 | 2 - 3 | 946 | 70 | |
GenBank mRNA | BT016046.1 | - | 2 - 3 | 917 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |