Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103491.1 | NP_001096961.1 | 11 - 12 | 2698 | 81 | |
NM_001298217.1 | NP_001285146.1 | 12 - 13 | 3078 | 81 | ||
NM_078581.3 | NP_511136.2 | 13 - 14 | 3458 | 81 | ||
GenBank mRNA | BT003214.1 | - | 13 - 14 | 3455 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |