Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298217.1 | NP_001285146.1 | 1 - 2 | 381 | 91 | |
NM_078581.3 | NP_511136.2 | 2 - 3 | 761 | 91 | ||
GenBank mRNA | BT003214.1 | - | 2 - 3 | 758 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |