Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103491.1 | NP_001096961.1 | 1 - 2 | 264 | 85 | |
NM_001298217.1 | NP_001285146.1 | 2 - 3 | 644 | 85 | ||
NM_078581.3 | NP_511136.2 | 3 - 4 | 1024 | 85 | ||
GenBank mRNA | BT003214.1 | - | 3 - 4 | 1021 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |