Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 7 - 8 | 2618 | 63 | |
NM_001144718.3 | NP_001138190.1 | 8 - 9 | 3323 | 63 | ||
NM_001169258.2 | NP_001162729.1 | 2 - 3 | 552 | 63 | ||
NM_001169259.3 | NP_001162730.2 | 8 - 9 | 2728 | 63 | ||
NM_001169260.3 | NP_001162731.1 | 9 - 10 | 3594 | 63 | ||
NM_001169261.3 | NP_001162732.2 | 7 - 8 | 2618 | 63 | ||
NM_001258711.2 | NP_001245640.1 | 5 - 6 | 1567 | 63 | ||
NM_001258712.2 | NP_001245641.1 | 5 - 6 | 1573 | 63 | ||
NM_001258713.2 | NP_001245642.1 | 5 - 6 | 1585 | 63 | ||
NM_001272554.2 | NP_001259483.1 | 5 - 6 | 1467 | 63 | ||
NM_078582.4 | NP_511137.3 | 3 - 4 | 753 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |