Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NetA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258738.2 | NP_001245667.1 | 2 - 3 | 1888 | 77 | |
NM_001298314.1 | NP_001285243.1 | 2 - 3 | 1982 | 77 | ||
NM_078599.5 | NP_511154.3 | 2 - 3 | 1888 | 77 | ||
GenBank mRNA | AY089580.1 | - | 2 - 3 | 1885 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |