Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042810.3 | NP_001036275.1 | 8 - 9 | 1567 | 65 | |
NM_001201662.2 | NP_001188591.1 | 7 - 8 | 1516 | 65 | ||
NM_001201663.2 | NP_001188592.1 | 8 - 9 | 1561 | 65 | ||
NM_001272627.1 | NP_001259556.1 | 7 - 8 | 1510 | 65 | ||
NM_001272628.1 | NP_001259557.1 | 7 - 8 | 1537 | 65 | ||
NM_001272629.1 | NP_001259558.1 | 7 - 8 | 1516 | 65 | ||
NM_078603.4 | NP_511158.2 | 8 - 9 | 1567 | 65 | ||
GenBank mRNA | BT150281.1 | - | 5 - 6 | 966 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |