Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042810.3 | NP_001036275.1 | 10 - 11 | 1814 | 109 | |
NM_001201662.2 | NP_001188591.1 | 9 - 10 | 1763 | 109 | ||
NM_001201663.2 | NP_001188592.1 | 10 - 11 | 1808 | 109 | ||
NM_001272627.1 | NP_001259556.1 | 9 - 10 | 1757 | 109 | ||
NM_001272628.1 | NP_001259557.1 | 9 - 10 | 1784 | 109 | ||
NM_001272629.1 | NP_001259558.1 | 9 - 10 | 1763 | 109 | ||
NM_078603.4 | NP_511158.2 | 10 - 11 | 1814 | 109 | ||
GenBank mRNA | BT150281.1 | - | 7 - 8 | 1213 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |