Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkcdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144719.2 | NP_001138191.1 | 4 - 5 | 1082 | 78 | |
NM_078616.4 | NP_511171.3 | 4 - 5 | 1258 | 78 | ||
GenBank mRNA | BT021275.1 | - | 3 - 4 | 1014 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |