Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kat80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298388.1 | NP_001285317.1 | 3 - 4 | 732 | 76 | |
NM_078639.3 | NP_523363.2 | 4 - 5 | 633 | 76 | ||
NM_167488.2 | NP_727940.1 | 3 - 4 | 732 | 76 | ||
GenBank mRNA | BT044203.1 | - | 3 - 4 | 713 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |