Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258777.1 | NP_001245706.1 | 3 - 4 | 3579 | 68 | |
NM_001258778.2 | NP_001245707.1 | 6 - 7 | 1394 | 68 | ||
NM_001298390.1 | NP_001285319.1 | 4 - 5 | 3527 | 68 | ||
NM_078640.4 | NP_523364.2 | 3 - 4 | 1147 | 68 | ||
NM_167489.2 | NP_727941.2 | 4 - 5 | 1283 | 68 | ||
NM_167490.3 | NP_727942.1 | 3 - 4 | 1215 | 68 | ||
NM_167491.2 | NP_727943.1 | 3 - 4 | 1347 | 68 | ||
NM_176741.3 | NP_788914.2 | 5 - 6 | 1353 | 68 | ||
GenBank mRNA | AY118437.1 | - | 3 - 4 | 1151 | 68 | |
L35604.1 | - | 3 - 4 | 1177 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |