Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078642.4 | NP_523366.2 | 1 - 2 | 290 | 73 | |
GenBank mRNA | BT031016.1 | - | 1 - 2 | 288 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |