Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01813733_m1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813733_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042816.2
NP_001036281.1
24 - 25
4397
65
NM_001201671.1
NP_001188600.1
24 - 25
4328
65
NM_001201672.1
NP_001188601.1
24 - 25
4328
65
NM_001201673.1
NP_001188602.1
23 - 24
4265
65
NM_001201674.1
NP_001188603.1
23 - 24
4295
65
NM_001201675.1
NP_001188604.1
23 - 24
4241
65
NM_001201676.1
NP_001188605.1
24 - 25
4304
65
NM_001201677.1
NP_001188606.1
24 - 25
4232
65
NM_001201678.1
NP_001188607.1
23 - 24
4265
65
NM_001201679.1
NP_001188608.1
24 - 25
4304
65
NM_001201680.1
NP_001188609.1
23 - 24
4304
65
NM_001201681.1
NP_001188610.1
23 - 24
4280
65
NM_001201682.1
NP_001188611.1
24 - 25
4367
65
NM_001201683.1
NP_001188612.1
24 - 25
4232
65
NM_001201684.1
NP_001188613.1
23 - 24
4241
65
NM_001201685.1
NP_001188614.1
23 - 24
4304
65
NM_001201686.1
NP_001188615.1
24 - 25
4334
65
NM_001201687.1
NP_001188616.1
24 - 25
4358
65
NM_001201688.1
NP_001188617.1
23 - 24
4295
65
NM_001201689.1
NP_001188618.1
23 - 24
4310
65
NM_001201690.1
NP_001188619.1
24 - 25
4379
65
NM_001201691.1
NP_001188620.1
24 - 25
4418
65
NM_001201692.1
NP_001188621.1
24 - 25
4271
65
NM_001201693.1
NP_001188622.1
24 - 25
4256
65
NM_001201694.1
NP_001188623.1
24 - 25
4262
65
NM_001201695.2
NP_001188624.1
24 - 25
4256
65
NM_001201698.1
NP_001188627.1
23 - 24
4334
65
NM_001201699.1
NP_001188628.1
22 - 23
4271
65
NM_001201700.1
NP_001188629.1
23 - 24
4325
65
NM_001201701.1
NP_001188630.1
22 - 23
4262
65
NM_001201702.1
NP_001188631.1
23 - 24
4400
65
NM_001201703.3
NP_001188632.1
24 - 25
4818
65
NM_001201704.1
NP_001188633.1
22 - 23
4337
65
NM_001201705.1
NP_001188634.1
23 - 24
4253
65
NM_001201706.1
NP_001188635.1
24 - 25
4433
65
NM_001201707.1
NP_001188636.1
24 - 25
4343
65
NM_001201709.1
NP_001188638.1
23 - 24
4271
65
NM_001201710.1
NP_001188639.1
20 - 21
4155
65
NM_001201711.1
NP_001188640.1
22 - 23
4205
65
NM_001201712.1
NP_001188641.1
21 - 22
4172
65
NM_001201713.1
NP_001188642.1
19 - 20
3513
65
NM_001201714.1
NP_001188643.1
19 - 20
3489
65
NM_001201715.1
NP_001188644.1
22 - 23
4190
65
NM_001201716.1
NP_001188645.1
23 - 24
4301
65
NM_001201719.1
NP_001188648.1
23 - 24
4400
65
NM_001272690.1
NP_001259619.1
24 - 25
4397
65
NM_001298403.1
NP_001285332.1
24 - 25
4550
65
NM_078647.4
NP_523371.2
24 - 25
4397
65
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0