Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01813736_m1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813736_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042815.2
NP_001036280.1
27 - 28
4840
148
NM_001042816.2
NP_001036281.1
27 - 28
4840
148
NM_001144733.2
NP_001138205.1
26 - 27
4789
148
NM_001201671.1
NP_001188600.1
27 - 28
4771
148
NM_001201672.1
NP_001188601.1
27 - 28
4771
148
NM_001201673.1
NP_001188602.1
26 - 27
4708
148
NM_001201674.1
NP_001188603.1
26 - 27
4738
148
NM_001201675.1
NP_001188604.1
26 - 27
4684
148
NM_001201676.1
NP_001188605.1
27 - 28
4747
148
NM_001201677.1
NP_001188606.1
27 - 28
4675
148
NM_001201678.1
NP_001188607.1
26 - 27
4708
148
NM_001201679.1
NP_001188608.1
27 - 28
4747
148
NM_001201680.1
NP_001188609.1
26 - 27
4747
148
NM_001201681.1
NP_001188610.1
26 - 27
4723
148
NM_001201682.1
NP_001188611.1
27 - 28
4810
148
NM_001201683.1
NP_001188612.1
27 - 28
4675
148
NM_001201684.1
NP_001188613.1
26 - 27
4684
148
NM_001201685.1
NP_001188614.1
26 - 27
4747
148
NM_001201686.1
NP_001188615.1
27 - 28
4777
148
NM_001201687.1
NP_001188616.1
27 - 28
4801
148
NM_001201688.1
NP_001188617.1
26 - 27
4738
148
NM_001201689.1
NP_001188618.1
26 - 27
4753
148
NM_001201690.1
NP_001188619.1
27 - 28
4822
148
NM_001201691.1
NP_001188620.1
27 - 28
4861
148
NM_001201692.1
NP_001188621.1
27 - 28
4714
148
NM_001201693.1
NP_001188622.1
27 - 28
4699
148
NM_001201694.1
NP_001188623.1
27 - 28
4705
148
NM_001201695.2
NP_001188624.1
27 - 28
4699
148
NM_001201696.1
NP_001188625.1
25 - 26
4705
148
NM_001201697.1
NP_001188626.1
27 - 28
4771
148
NM_001201698.1
NP_001188627.1
26 - 27
4777
148
NM_001201699.1
NP_001188628.1
25 - 26
4714
148
NM_001201700.1
NP_001188629.1
26 - 27
4768
148
NM_001201701.1
NP_001188630.1
25 - 26
4705
148
NM_001201702.1
NP_001188631.1
26 - 27
4843
148
NM_001201703.3
NP_001188632.1
27 - 28
5261
148
NM_001201704.1
NP_001188633.1
25 - 26
4780
148
NM_001201705.1
NP_001188634.1
26 - 27
4696
148
NM_001201706.1
NP_001188635.1
27 - 28
4876
148
NM_001201707.1
NP_001188636.1
27 - 28
4786
148
NM_001201708.1
NP_001188637.1
26 - 27
4738
148
NM_001201709.1
NP_001188638.1
26 - 27
4714
148
NM_001201710.1
NP_001188639.1
23 - 24
4598
148
NM_001201711.1
NP_001188640.1
25 - 26
4648
148
NM_001201712.1
NP_001188641.1
24 - 25
4615
148
NM_001201713.1
NP_001188642.1
22 - 23
3956
148
NM_001201714.1
NP_001188643.1
22 - 23
3932
148
NM_001201715.1
NP_001188644.1
25 - 26
4633
148
NM_001201716.1
NP_001188645.1
26 - 27
4744
148
NM_001201717.1
NP_001188646.1
26 - 27
4843
148
NM_001201718.1
NP_001188647.1
25 - 26
4780
148
NM_001201719.1
NP_001188648.1
26 - 27
4843
148
NM_001201720.1
NP_001188649.1
25 - 26
4645
148
NM_001201721.1
NP_001188650.1
21 - 22
3703
148
NM_001201722.1
NP_001188651.1
25 - 26
4615
148
NM_001201723.1
NP_001188652.1
25 - 26
4656
148
NM_001272690.1
NP_001259619.1
27 - 28
4840
148
NM_001298403.1
NP_001285332.1
27 - 28
4993
148
NM_001298404.1
NP_001285333.1
25 - 26
4612
148
NM_078647.4
NP_523371.2
27 - 28
4840
148
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0