Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B-H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272713.1 | NP_001259642.1 | 2 - 3 | 1275 | 67 | |
NM_078663.3 | NP_523387.1 | 2 - 3 | 1270 | 67 | ||
GenBank mRNA | AY058309.1 | - | 2 - 3 | 1247 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |