Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beta-Spec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078664.3 | NP_523388.1 | 4 - 5 | 6781 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |