Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKLR-17D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103553.3 | NP_001097023.1 | 1 - 2 | 816 | 63 | |
GenBank mRNA | AY231149.1 | - | 1 - 2 | 1020 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |