Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169321.2 | NP_001162792.1 | 3 - 4 | 602 | 63 | |
NM_001298486.1 | NP_001285415.1 | 3 - 4 | 694 | 63 | ||
NM_078683.3 | NP_523407.1 | 3 - 4 | 694 | 63 | ||
GenBank mRNA | AJ401391.1 | - | 3 - 4 | 580 | 63 | |
AY051751.1 | - | 3 - 4 | 672 | 63 | ||
AY079186.1 | - | 3 - 4 | 581 | 63 | ||
AY079188.1 | - | 3 - 4 | 614 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |