Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078684.4 | NP_523408.2 | 9 - 10 | 4871 | 66 | |
GenBank mRNA | AY051675.1 | - | 9 - 10 | 4811 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |