Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp19d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078697.3 | NP_523421.2 | 1 - 2 | 669 | 67 | |
NM_176767.2 | NP_788940.1 | 1 - 2 | 192 | 67 | ||
GenBank mRNA | AY118484.1 | - | 2 - 3 | 688 | 67 | |
KX532139.1 | - | 1 - 2 | 144 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |