Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298588.1 | NP_001285517.1 | 1 - 2 | 163 | 81 | |
NM_078710.4 | NP_523434.2 | 1 - 2 | 163 | 81 | ||
GenBank mRNA | AY070624.1 | - | 1 - 2 | 154 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |