Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144289.2 | NP_001137761.1 | 12 - 13 | 13014 | 58 | |
NM_001258889.1 | NP_001245818.1 | 14 - 15 | 12492 | 58 | ||
NM_001258890.1 | NP_001245819.1 | 12 - 13 | 12219 | 58 | ||
NM_001258891.2 | NP_001245820.1 | 12 - 13 | 12217 | 58 | ||
NM_078717.3 | NP_523441.1 | 13 - 14 | 12429 | 58 | ||
NM_164376.2 | NP_722617.1 | 2 - 3 | 2711 | 58 | ||
GenBank mRNA | AF215703.1 | - | 13 - 14 | 12401 | 58 | |
MN044634.1 | - | 13 - 14 | 12474 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |