Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144289.2 | NP_001137761.1 | 3 - 4 | 8477 | 101 | |
NM_001258889.1 | NP_001245818.1 | 3 - 4 | 8477 | 101 | ||
NM_001258890.1 | NP_001245819.1 | 3 - 4 | 8477 | 101 | ||
NM_001258891.2 | NP_001245820.1 | 3 - 4 | 8658 | 101 | ||
NM_078717.3 | NP_523441.1 | 3 - 4 | 8477 | 101 | ||
GenBank mRNA | AF215703.1 | - | 3 - 4 | 8449 | 101 | |
MN044634.1 | - | 3 - 4 | 8522 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |