Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078720.3 | NP_523444.1 | 1 - 2 | 324 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |