Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272996.1 | NP_001259925.1 | 4 - 5 | 846 | 66 | |
NM_001272997.1 | NP_001259926.1 | 4 - 5 | 837 | 66 | ||
NM_001272998.1 | NP_001259927.1 | 4 - 5 | 836 | 66 | ||
NM_078733.4 | NP_523457.1 | 4 - 5 | 819 | 66 | ||
NM_164475.2 | NP_722795.1 | 4 - 5 | 826 | 66 | ||
NM_164476.2 | NP_722796.1 | 4 - 5 | 932 | 66 | ||
NM_164477.3 | NP_722797.1 | 4 - 5 | 1128 | 66 | ||
NM_164478.2 | NP_722798.1 | 4 - 5 | 800 | 66 | ||
NM_164479.2 | NP_722799.1 | 4 - 5 | 784 | 66 | ||
GenBank mRNA | AB035353.1 | - | 4 - 5 | 834 | 66 | |
AB035671.1 | - | 4 - 5 | 805 | 66 | ||
AY060343.1 | - | 4 - 5 | 1104 | 66 | ||
AY081180.1 | - | 3 - 4 | 704 | 66 | ||
AY081181.1 | - | 4 - 5 | 788 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |