Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: msl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078743.5 | NP_523467.1 | 1 - 2 | 2407 | 67 | |
NM_164536.2 | NP_722907.1 | 2 - 3 | 2274 | 67 | ||
GenBank mRNA | AY128426.1 | - | 2 - 3 | 2257 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |