Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ed | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273084.1 | NP_001260013.1 | 6 - 7 | 3376 | 71 | |
NM_001273085.1 | NP_001260014.1 | 6 - 7 | 3376 | 71 | ||
NM_078745.3 | NP_523469.2 | 6 - 7 | 3376 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |