Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103608.2 | NP_001097078.1 | 4 - 5 | 2311 | 66 | |
NM_001103609.3 | NP_001097079.1 | 4 - 5 | 2648 | 66 | ||
NM_078747.3 | NP_523471.2 | 4 - 5 | 2648 | 66 | ||
NM_164580.2 | NP_722971.1 | 4 - 5 | 2313 | 66 | ||
GenBank mRNA | BT029941.1 | - | 4 - 5 | 2311 | 66 | |
BT029944.1 | - | 4 - 5 | 2311 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |