Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014467.2 | NP_001014467.1 | 3 - 4 | 1024 | 63 | |
NM_001014468.2 | NP_001014468.1 | 2 - 3 | 588 | 63 | ||
NM_001014469.2 | NP_001014469.1 | 3 - 4 | 1024 | 63 | ||
NM_001201761.2 | NP_001188690.1 | 2 - 3 | 588 | 63 | ||
NM_001273154.2 | NP_001260083.1 | 2 - 3 | 438 | 63 | ||
NM_001273155.1 | NP_001260084.1 | 2 - 3 | 617 | 63 | ||
NM_001298682.1 | NP_001285611.1 | 2 - 3 | 534 | 63 | ||
NM_001298683.1 | NP_001285612.1 | 2 - 3 | 438 | 63 | ||
NM_078756.4 | NP_523480.2 | 3 - 4 | 1024 | 63 | ||
NM_164625.2 | NP_723057.1 | 2 - 3 | 588 | 63 | ||
NM_164626.3 | NP_723058.2 | 2 - 3 | 534 | 63 | ||
NM_164627.2 | NP_723059.2 | 2 - 3 | 617 | 63 | ||
GenBank mRNA | AY058777.1 | - | 3 - 4 | 1007 | 63 | |
BT100049.1 | - | 2 - 3 | 557 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |