Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014469.2 | NP_001014469.1 | 6 - 7 | 2133 | 70 | |
NM_001273154.2 | NP_001260083.1 | 5 - 6 | 1547 | 70 | ||
NM_078756.4 | NP_523480.2 | 6 - 7 | 2133 | 70 | ||
NM_164625.2 | NP_723057.1 | 5 - 6 | 1697 | 70 | ||
NM_164626.3 | NP_723058.2 | 5 - 6 | 1643 | 70 | ||
NM_164627.2 | NP_723059.2 | 5 - 6 | 1726 | 70 | ||
GenBank mRNA | AY058777.1 | - | 6 - 7 | 2116 | 70 | |
BT100049.1 | - | 5 - 6 | 1666 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |