Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nompC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258962.3 | NP_001245891.1 | 12 - 13 | 5124 | 90 | |
NM_205912.4 | NP_995634.2 | 12 - 13 | 5124 | 90 | ||
GenBank mRNA | HM582118.1 | - | 12 - 13 | 4466 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster Inventoried |