Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103637.3 | NP_001097107.1 | 4 - 5 | 2592 | 64 | |
NM_001103638.3 | NP_001097108.1 | 4 - 5 | 1187 | 64 | ||
NM_001103639.4 | NP_001097109.2 | 3 - 4 | 1690 | 64 | ||
NM_001298777.1 | NP_001285706.1 | 3 - 4 | 2553 | 64 | ||
GenBank mRNA | BT050481.1 | - | 4 - 5 | 1598 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |