Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CdGAPr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169546.2 | NP_001163017.1 | 5 - 6 | 3731 | 77 | |
NM_001169547.2 | NP_001163018.1 | 5 - 6 | 3731 | 77 | ||
NM_001273671.2 | NP_001260600.1 | 6 - 7 | 3806 | 77 | ||
NM_136158.5 | NP_610002.1 | 5 - 6 | 3734 | 77 | ||
GenBank mRNA | AF218776.1 | - | 5 - 6 | 3723 | 77 | |
BT132944.1 | - | 5 - 6 | 3701 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |